2-{[(4-Fluorophenyl)sulphonyl]amino}-3-methylbutanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Fluorobenzenesulphamido)-3-methylbutyric acid

PC9372-1G PC9372-5G
APOSPC9372-1GEA 183 EUR
APOSPC9372-1G APOSPC9372-5G
2-{[(4-Fluorophenyl)sulphonyl]amino}-3-methylbutanoic acid 97%
2-{[(4-Fluorophenyl)sulphonyl]amino}-3-methylbutanoic acid
Formel: C₁₁H₁₄FNO₄S
Molecular Weight: 275.3 g/mol
Schmelzpunkt: 159…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664351
CAS-Nummer: 250714-79-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR