2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-Fluorobenzenesulphamido)-2-phenylacetic acid

PC9374-500MG
APOSPC9374-500MGEA 21.9 EUR
APOSPC9374-500MG
2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid 97%
2-{[(4-Fluorophenyl)sulphonyl]amino}-2-phenylacetic acid
Formel: C₁₄H₁₂FNO₄S
Molecular Weight: 309,32 g/mol
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173006
CAS-Nummer: 117309-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR