2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide

Lieferant: Apollo Scientific

PC9382-1G PC9382-500MG PC9382-5G
APOSPC9382-1GEA 275 EUR
APOSPC9382-1G APOSPC9382-500MG APOSPC9382-5G
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide
2-Hydrazino-2-oxo-N-[4-(trifluoromethoxy)phenyl]acetamide
Formel: C₉H₈F₃N₃O₃
Schmelzpunkt: 215…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173093
CAS-Nummer: 338405-24-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR