4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

PC9384-1G
APOSPC9384-1GEA 569 EUR
APOSPC9384-1G
4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid 97%
4-Hydroxy-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylic acid
Formel: C₁₂H₇F₃N₂O₄
Molecular Weight: 300,2 g/mol
Schmelzpunkt: 261…263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00141002
CAS-Nummer: 121582-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR