5-Fluoro-1H-benzimidazole 98%

Lieferant: Apollo Scientific

Synonyme: 5-Fluoro-1H-benzo[d]imidazole

PC9393-5G
APOSPC9393-5GEA 74.1 EUR
APOSPC9393-5G
5-Fluoro-1H-benzimidazole 98%
5-Fluoro-1H-benzimidazole
Formel: C₇H₅FN₂
Molecular Weight: 136.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00792438
CAS-Nummer: 1977-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR