Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate 95%

Lieferant: Apollo Scientific
Achtung

PC9397-1G
APOSPC9397-1GEA 140 EUR
APOSPC9397-1G
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate 95%
Methyl-3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxylate
Formel: C₁₂H₉ClFNO₃
Molecular Weight: 269,66 g/mol
Schmelzpunkt: 53…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00214743
CAS-Nummer: 4415-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR