Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Lieferant: Apollo Scientific
PC9401-500G
APOSPC9401-500GEA
590
EUR
APOSPC9401-500G
Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Methyl 3-{[2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-methylhydrazino]carbonyl}isonicotinate
Formel:
C₁₅H₁₂ClF₃N₄O₃ Siedepunkt: 168…170 °C Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00975038 CAS-Nummer: 338415-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...