Methyl-3-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-thiophenecarboxylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9402-1G
APOSPC9402-1GEA
178
EUR
APOSPC9402-1G
Methyl-3-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-thiophenecarboxylate 97%
Methyl-3-{[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}-2-thiophenecarboxylate
Formel:
C₁₂H₇ClF₃NO₃S Molecular Weight: 337,71 g/mol Schmelzpunkt: 63…64 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00141796 CAS-Nummer: 338756-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...