Methyl 1-[2,6-dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate 97%

Lieferant: Apollo Scientific

PC9406-1G
APOSPC9406-1GEA 977 EUR
APOSPC9406-1G
Methyl 1-[2,6-dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate 97%
Methyl 1-[2,6-dinitro-4-(trifluoromethyl)phenyl]piperidine-4-carboxylate
Formel: C₁₄H₁₄F₃N₃O₆
Molecular Weight: 377,273 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00664468
CAS-Nummer: 303144-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR