Methyl 3-(4-fluoro-2-nitrophenoxy)thiophene-2-carboxylate 97%

Lieferant: Apollo Scientific

PC9413-1G
APOSPC9413-1GEA 402 EUR
APOSPC9413-1G
Methyl 3-(4-fluoro-2-nitrophenoxy)thiophene-2-carboxylate 97%
Methyl 3-(4-fluoro-2-nitrophenoxy)thiophene-2-carboxylate
Formel: C₁₂H₈FNO₅S
Molecular Weight: 297,259 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00214991
CAS-Nummer: 303152-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR