6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydropyridine-3-carbonitrile
PC9432-1G
APOSPC9432-1GEA
58
EUR
APOSPC9432-1G
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
6-Methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydro-3-pyridinecarbonitrile
Formel:
C₈H₅F₃N₂O Molecular Weight: 202,14 g/mol Schmelzpunkt: 232…234 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01314908 CAS-Nummer: 654-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...