5-Methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-5-methylisoxazole-4-carbox(3-trifluoromethyl)anilide

PC9441-500MG
APOSPC9441-500MGEA 415 EUR
APOSPC9441-500MG
5-Methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylic acid
5-Methyl-4-{[3-(trifluoromethyl)anilino]carbonyl}-3-isoxazolecarboxylic acid
Formel: C₁₃H₉F₃N₂O₄
Molecular Weight: 314,22 g/mol
Schmelzpunkt: 178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00793091
CAS-Nummer: 339012-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR