6-(Trifluoromethyl)quinoline-4-thiol

Lieferant: Apollo Scientific
Achtung

PC9444-1G PC9444-500MG
APOSPC9444-1GEA 667 EUR
APOSPC9444-1G APOSPC9444-500MG
6-(Trifluoromethyl)quinoline-4-thiol
6-(Trifluoromethyl)quinoline-4-thiol
Formel: C₁₀H₆F₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD00269611
CAS-Nummer: 544716-08-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR