1-Methyl-2-[3-(trifluoromethyl)phenylthio]indole-3-carboxaldehyde

Lieferant: Apollo Scientific

PC9449-100MG
APOSPC9449-100MGEA 271 EUR
APOSPC9449-100MG
1-Methyl-2-[3-(trifluoromethyl)phenylthio]indole-3-carboxaldehyde
1-Methyl-2-[3-(trifluoromethyl)phenylthio]indole-3-carboxaldehyde
Formel: C₁₇H₁₂F₃NOS
Schmelzpunkt: 165…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01568347
CAS-Nummer: 338416-52-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR