5-Fluor-3-methylbenzo[b]thiophen 97%

Lieferant: Apollo Scientific
Achtung

PC9452-1G
APOSPC9452-1GEA 29 EUR
APOSPC9452-1G
5-Fluor-3-methylbenzo[b]thiophen 97%
5-Fluor-3-methylbenzo[b]thiophen
Formel: C₉H₇FS
Siedepunkt: 68 °C/0,5mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00179348
CAS-Nummer: 17514-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR