2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid 97%
Lieferant: Apollo Scientific
Synonyme:
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylglycine
PC9482-1G
PC9482-500MG
PC9482-5G
APOSPC9482-1GEA
275
EUR
APOSPC9482-1G
APOSPC9482-500MG
APOSPC9482-5G
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid 97%
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)acetic acid
Formel:
C₁₅H₁₂F₃NO₄S Schmelzpunkt: 169…171 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172802 CAS-Nummer: 250714-41-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...