N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid

PC9484-1G
APOSPC9484-1GEA 116 EUR
APOSPC9484-1G
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine 97%
N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylalanine
Formel: C₁₆H₁₄F₃NO₄S
Molecular Weight: 373,35 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173005
CAS-Nummer: 250714-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR