5-(Hydroxymethyl)-1-{2-[3-(trifluoromethyl)phenoxy]ethyl}-1H-imidazole-2-thiol 97%

Lieferant: Apollo Scientific

Synonyme: 5-(Hydroxymethyl)-2-mercapto-1-{2-[3-(trifluoromethyl)phenoxy]ethyl}-1H-imidazole

PC9486-100MG
APOSPC9486-100MGEA 271 EUR
APOSPC9486-100MG
5-(Hydroxymethyl)-1-{2-[3-(trifluoromethyl)phenoxy]ethyl}-1H-imidazole-2-thiol 97%
5-(Hydroxymethyl)-1-{2-[3-(trifluoromethyl)phenoxy]ethyl}-1H-imidazole-2-thiol
Formel: C₁₃H₁₃F₃N₂O₂S
Molecular Weight: 318,315 g/mol
Schmelzpunkt: 209…211 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215162
CAS-Nummer: 338794-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR