5-(4-tert-Butylbenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde

Lieferant: Apollo Scientific

PC9488-100MG
APOSPC9488-100MGEA 271 EUR
APOSPC9488-100MG
5-(4-tert-Butylbenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde
5-(4-tert-Butylbenzylthio)-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxaldehyde
Formel: C₁₇H₁₉F₃N₂OS
Molecular Weight: 356,406 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140860
CAS-Nummer: 321553-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR