3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-2-thiophenecarboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-[3-(Trifluoromethyl)benzylthio]thiophene-2-carboxylic acid

PC9491-1G
APOSPC9491-1GEA 268 EUR
APOSPC9491-1G
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-2-thiophenecarboxylic acid 97%
3-{[3-(Trifluoromethyl)benzyl]sulphanyl}-2-thiophenecarboxylic acid
Formel: C₁₃H₉F₃O₂S₂
Molecular Weight: 318,34 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00663855
CAS-Nummer: 251096-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR