4-Chloro-3-fluorophenethyl alcohol 95+%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Chloro-3-fluorophenyl)ethanol

PC9496-1G
APOSPC9496-1GEA 119 EUR
APOSPC9496-1G
4-Chloro-3-fluorophenethyl alcohol 95+%
4-Chloro-3-fluorophenethyl alcohol
Formel: C₈H₈ClFO
Molecular Weight: 174,6 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06201151
CAS-Nummer: 206362-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR