2-({[3-(Trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC9503-1G
APOSPC9503-1GEA 174 EUR
APOSPC9503-1G
2-({[3-(Trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid 97%
2-({[3-(Trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid
Formel: C₁₀H₁₀F₃NO₄S
Schmelzpunkt: 123…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00172805
CAS-Nummer: 288266-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR