2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC9506-50MG
APOSPC9506-50MGEA
14.7
EUR
APOSPC9506-50MG
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
2-[3-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid
Formel:
C₁₁H₆F₃NO₂S Schmelzpunkt: 200…202 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00142039 CAS-Nummer: 115311-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...