1-Bromo-2-(1,1,2,2-tetrafluoroethoxy)benzene

Lieferant: Apollo Scientific
Achtung

PC9512-1G
APOSPC9512-1GEA 27.7 EUR
APOSPC9512-1G
1-Bromo-2-(1,1,2,2-tetrafluoroethoxy)benzene
1-Bromo-2-(1,1,2,2-tetrafluoroethoxy)benzene
Formel: C₈H₅BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD03094557
CAS-Nummer: 883499-00-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR