(4-Chlor-2-fluor-3-methyl-6-(trifluormethyl)phenyl)essigsäure 98%

Lieferant: Apollo Scientific

PC9514-1G
APOSPC9514-1GEA 945 EUR
APOSPC9514-1G
(4-Chlor-2-fluor-3-methyl-6-(trifluormethyl)phenyl)essigsäure 98%
(4-Chlor-2-fluor-3-methyl-6-(trifluormethyl)phenyl)essigsäure
Formel: C₁₀H₇ClF₄O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 914635-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR