4-Fluorobenzylhydrazine hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: (4-Fluorobenzyl)hydrazine hydrochloride

PC9518-1G
APOSPC9518-1GEA 81.2 EUR
APOSPC9518-1G
4-Fluorobenzylhydrazine hydrochloride
4-Fluorobenzylhydrazine hydrochloride
Formel: C₇H₁₀ClFN₂
Molecular Weight: 176,62 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08458859
CAS-Nummer: 1059626-05-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR