4-(Difluoromethoxy)benzylhydrazine hydrochloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: Difluoromethyl 4-(hydrazinomethyl)phenyl ether hydrochloride, 1-(Difluoromethoxy)-4-(hydrazinomethyl)benzene hydrochloride

PC9519-1G
APOSPC9519-1GEA 0 EUR
APOSPC9519-1G
4-(Difluoromethoxy)benzylhydrazine hydrochloride
4-(Difluoromethoxy)benzylhydrazine hydrochloride
Formel: C₈H₁₀F₂N₂O·ClH
Molecular Weight: 224.636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08458142
CAS-Nummer: 887596-65-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR