Ethyl(3,4-difluorbenzoyl)formiat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl (3,4-difluorophenyl)(oxo)acetate, Ethyl 2-(3,4-difluorophenyl)-2-oxoacetate

PC9522-100MG
APOSPC9522-100MGEA 21.9 EUR
APOSPC9522-100MG
Ethyl(3,4-difluorbenzoyl)formiat
Ethyl(3,4-difluorbenzoyl)formiat
Formel: C₁₀H₈F₂O₃
Molecular Weight: 214,1655 g/mol
Siedepunkt: 105 °C/0,1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01319619
CAS-Nummer: 73790-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR