2,6-Dimethyl-4-fluorochlorobenzene 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-fluoro-m-xylene, 2-Chloro-1,3-dimethyl-5-fluorobenzene

PC9538-1G
APOSPC9538-1GEA 655 EUR
APOSPC9538-1G
2,6-Dimethyl-4-fluorochlorobenzene 98%
2,6-Dimethyl-4-fluorochlorobenzene
Formel: C₈H₈ClF
Molecular Weight: 158,601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972723
CAS-Nummer: 14994-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR