4-Chloro-2-fluoro-3-methyl-6-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific

PC9540-1G
APOSPC9540-1GEA 851 EUR
APOSPC9540-1G
4-Chloro-2-fluoro-3-methyl-6-(trifluoromethyl)aniline 98%
4-Chloro-2-fluoro-3-methyl-6-(trifluoromethyl)aniline
Formel: C₈H₆ClF₄N
Molecular Weight: 227,587 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701045
CAS-Nummer: 914635-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR