3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC9567-1G
PC9567-5G
PC9567-10G
APOSPC9567-1GEA
348
EUR
APOSPC9567-1G
APOSPC9567-5G
APOSPC9567-10G
3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%
3-Isocyanato-1-(trifluoroacetyl)piperidine
- May form small amount of sediment.
Formel:
C₈H₉F₃N₂O₂ Molecular Weight: 222.17 g/mol Siedepunkt: 129…132 °C/5.25mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424489 CAS-Nummer: 808764-41-2 UN: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...