3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%

Lieferant: Apollo Scientific
Gefahr

PC9567-1G PC9567-5G PC9567-10G
APOSPC9567-1GEA 348 EUR
APOSPC9567-1G APOSPC9567-5G APOSPC9567-10G
3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%
3-Isocyanato-1-(trifluoroacetyl)piperidine

  • May form small amount of sediment.

Formel: C₈H₉F₃N₂O₂
Molecular Weight: 222.17 g/mol
Siedepunkt: 129…132 °C/5.25mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03424489
CAS-Nummer: 808764-41-2
UN: 2206
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR