3-(Trifluoromethoxy)phenylacetylene 98%

Lieferant: Apollo Scientific
Achtung

PC9568-1G PC9568-5G PC9568-250MG
APOSPC9568-1GEA 0 EUR
APOSPC9568-1G APOSPC9568-5G APOSPC9568-250MG
3-(Trifluoromethoxy)phenylacetylene 98%
3-(Trifluoromethoxy)phenylacetylene
Formel: C₉H₅F₃O
Lagertemperatur: Kühlschrank
MDL: MFCD08458148
CAS-Nummer: 866683-57-0
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR