4-Brom-3-fluorbenzonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC9579-100G
PC9579-25G
PC9579-5G
APOSPC9579-100GEA
241
EUR
APOSPC9579-100G
APOSPC9579-25G
APOSPC9579-5G
4-Brom-3-fluorbenzonitril 95%
4-Brom-3-fluorbenzonitril
Formel:
C₇H₃BrFN Molecular Weight: 200.01 g/mol Schmelzpunkt: 92…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782069 CAS-Nummer: 133059-44-6 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...