1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(6-Trifluoromethyl-2-pyridyl)-1,4-diazepane
PC9582-100MG
APOSPC9582-100MGEA
111
EUR
APOSPC9582-100MG
1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
1-[3-(Trifluoromethyl)pyridin-2-yl]-1,4-diazepane
Formel:
C₁₁H₁₄F₃N₃ Molecular Weight: 245,249 g/mol Siedepunkt: 90 °C/0,2mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756231 CAS-Nummer: 243666-15-1 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...