2-Amino-5-fluorobenzeneboronic acid, neopentyl glycol ester, N-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[(tert-Butoxycarbonyl)amino]-5-fluorobenzeneboronic acid neopentyl glycol ester, tert-Butyl [2-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)-4-fluorophenyl]carbamate
PC9589-50MG
APOSPC9589-50MGEA
40.6
EUR
APOSPC9589-50MG
2-Amino-5-fluorobenzeneboronic acid, neopentyl glycol ester, N-BOC protected
2-Amino-5-fluorobenzeneboronic acid, neopentyl glycol ester, N-BOC protected
Formel:
C₁₆H₂₃BFNO₄ Molecular Weight: 323,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07776961 CAS-Nummer: 1218791-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...