5-Fluor-2-iodtoluol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluor-6-iodtoluol, 1-Fluor-4-iod-3-methylbenzol

PC9595-500G
APOSPC9595-500GEA 246 EUR
APOSPC9595-500G
5-Fluor-2-iodtoluol
5-Fluor-2-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236.03 g/mol
Siedepunkt: 138…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039397
CAS-Nummer: 66256-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR