3-(Chlormethyl)-5-(4-(trifluormethyl)phenyl)-1,2,4-oxadiazol 95+%
Lieferant: Apollo Scientific
PC9600-1G
APOSPC9600-1GEA
183
EUR
APOSPC9600-1G
3-(Chlormethyl)-5-(4-(trifluormethyl)phenyl)-1,2,4-oxadiazol 95+%
3-(Chloromethyl)-5-(4-(trifluoromethyl)phenyl)-1,2,4-oxadiazole
Formel:
C₁₀H₆ClF₃N₂O Schmelzpunkt: 48…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173952 CAS-Nummer: 175205-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...