Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9642-25G
PC9642-5G
APOSPC9642-25GEA
306
EUR
APOSPC9642-25G
APOSPC9642-5G
Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen 97%
Trifluor(1,1,2,2-tetrafluor-2-bromethoxy)ethylen
Formel:
F₂CCFOCF₂CF₂Br Siedepunkt: 55 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02183517 CAS-Nummer: 85737-06-0 EINECS: 288-516-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...