2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole, 4-(4-Formyl-1,3-thiazol-2-yl)benzotrifluoride
PC9656-1G
APOSPC9656-1GEA
190
EUR
APOSPC9656-1G
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
2-[4-(Trifluoromethyl)phenyl]-1,3-thiazole-4-carboxaldehyde
Formel:
C₁₁H₆F₃NOS Schmelzpunkt: 103…105 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07772870 CAS-Nummer: 174006-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...