Tetrafluorobenzene-1,2-diol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4,5,6-Tetrafluorobenzene-1,2-diol

PC9657-250MG
APOSPC9657-250MGEA 335 EUR
APOSPC9657-250MG
Tetrafluorobenzene-1,2-diol
Tetrafluorobenzene-1,2-diol
Formel: C₆H₂F₄O₂
Molecular Weight: 182,08 g/mol
Schmelzpunkt: 68…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075279
CAS-Nummer: 1996-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR