2-Chlor-3-fluorbenzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9671-5G
PC9671-25G
APOSPC9671-5GEA
33.5
EUR
APOSPC9671-5G
APOSPC9671-25G
2-Chlor-3-fluorbenzonitril 97%
2-Chlor-3-fluorbenzonitril
Formel:
C₇H₃ClFN Schmelzpunkt: 55…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407966 CAS-Nummer: 874781-08-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...