1-(4-Fluorphenyl)-2-pyrrolidinon 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Fluorophenyl)pyrrolidin-2-one

PC9696-1G PC9696-5G PC9696-250MG
APOSPC9696-1GEA 94.5 EUR
APOSPC9696-1G APOSPC9696-5G APOSPC9696-250MG
1-(4-Fluorphenyl)-2-pyrrolidinon 95+%
1-(4-Fluorphenyl)-2-pyrrolidinon
Formel: C₁₀H₁₀FNO
Molecular Weight: 179.19 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00138504
CAS-Nummer: 54660-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR