1-Chloro-2,4-dinitro-5-fluorobenzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC9715-1G
APOSPC9715-1GEA
524
EUR
APOSPC9715-1G
1-Chloro-2,4-dinitro-5-fluorobenzene 98%
1-Chloro-2,4-dinitro-5-fluorobenzene
Formel:
C₆H₂ClFN₂O₄ Molecular Weight: 220,542 g/mol Schmelzpunkt: 74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047671 CAS-Nummer: 327-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...