SpectraSynQ 1621
Lieferant: Apollo Scientific
|
Achtung
|
PC9725-1G
APOSPC9725-1GEA
101
EUR
APOSPC9725-1G
SpectraSynQ 1621
SpectraSynQ 1621
Fluorinated phosphazene for MS, especially suitable for +ve and -ve FAB high resolution MS (Jiang & Moini, ASMS Poster 1992), showing peaks upto 2120 mass units. R= -CH2(CF2CF2)xH, x=1,2 or 3.
Siedepunkt:
160…220 °C/0.01mmHg Dichte: 1.8 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08461619 CAS-Nummer: 105809-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...