2,2-Difluoro-2-phenylacetamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9727-1G
PC9727-5G
APOSPC9727-1GEA
67.2
EUR
APOSPC9727-1G
APOSPC9727-5G
2,2-Difluoro-2-phenylacetamide 97%
2,2-Difluoro-2-phenylacetamide
Formel:
C₈H₇F₂NO Molecular Weight: 171.144 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00183287 CAS-Nummer: 383-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...