2,2-Difluoro-2-phenylacetamide 97%

Lieferant: Apollo Scientific
Achtung

PC9727-1G PC9727-5G
APOSPC9727-1GEA 67.2 EUR
APOSPC9727-1G APOSPC9727-5G
2,2-Difluoro-2-phenylacetamide 97%
2,2-Difluoro-2-phenylacetamide
Formel: C₈H₇F₂NO
Molecular Weight: 171.144 g/mol
Schmelzpunkt: 114…115 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00183287
CAS-Nummer: 383-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR