6-Fluor-3-indolylessigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluorindol-3-ylessigsäure, 6-Fluorindol-3-essigsäure

PC9731-1G PC9731-500MG PC9731-5G
APOSPC9731-1GEA 71.5 EUR
APOSPC9731-1G APOSPC9731-500MG APOSPC9731-5G
6-Fluor-3-indolylessigsäure
6-Fluor-3-indolylessigsäure
Formel: C₁₀H₈FNO₂
Molecular Weight: 193.18 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074503
CAS-Nummer: 443-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR