3-Chlor-2,6-difluoranisol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-2,4-difluoro-3-methoxybenzene

PC9732-1G
APOSPC9732-1GEA 101 EUR
APOSPC9732-1G
3-Chlor-2,6-difluoranisol 98%
3-Chlor-2,6-difluoranisol
Formel: C₇H₅ClF₂O
Molecular Weight: 178.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631331
CAS-Nummer: 261762-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR