3,5-Difluor-2-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

PC9743-1G PC9743-5G PC9743-10G
APOSPC9743-1GEA 29 EUR
APOSPC9743-1G APOSPC9743-5G APOSPC9743-10G
3,5-Difluor-2-nitrophenol 98%
3,5-Difluor-2-nitrophenol
Formel: C₆H₃F₂NO₃
Molecular Weight: 175.0900 g/mol
Schmelzpunkt: 46…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08458165
CAS-Nummer: 151414-46-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR